Key Documents
G5920
Sigma-Aldrich
GW4716
≥98% (HPLC), solid
Synonym(s):
4-Hydroxy-benzoic acid(4-diethylamino-benzylidene)-hydrazide, GSK4716
Sign Into View Organizational & Contract Pricing
About This Item
UNSPSC Code:
51111800
PubChem Substance ID:
assay
≥98% (HPLC)
form
solid
color
yellow
solubility
DMSO: ≥14 mg/mL
H2O: insoluble
storage temp.
2-8°C
Gene Information
human ... ESRRG(2104)
Biochem/physiol Actions
GW4716 is an estrogen-related receptor γ (ERRγ) agonist.
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)